The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138510.1 | NP_612519.1 | 8 - 9 | 975 | 123 |
GenBank mRNA | BC086579.1 | - | 8 - 9 | 974 | 123 |
BC100248.1 | - | 8 - 9 | 969 | 123 | |
D14424.1 | - | 8 - 9 | 960 | 123 | |
L32601.1 | - | 8 - 9 | 975 | 123 |