Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1c12
Entrez Gene ID
361266
Gene Name
aldo-keto reductase family 1, member C12
Gene Aliases
-
Chromosome Location
Chr.17: 69614486 - 69647772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170342.1 NP_001163813.1 8 - 9 972 154
XM_006254190.2 XP_006254252.1 10 - 11 1822 154
XM_006254191.1 XP_006254253.1 11 - 12 1328 154
XM_008771873.1 XP_008770095.1 10 - 11 1212 154
XM_008771874.1 XP_008770096.1 10 - 11 1138 154
GenBank mRNA CN542908.1 - 1 - 2 154

View Full Product Details