Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acbd7
Entrez Gene ID
361277
Gene Name
acyl-CoA binding domain containing 7
Gene Aliases
RGD1564164
Chromosome Location
Chr.17: 78905338 - 78912042 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126079.1 NP_001119551.1 2 - 3 175 83

View Full Product Details