Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd16
Entrez Gene ID
307102
Gene Name
ankyrin repeat domain 16
Gene Aliases
-
Chromosome Location
Chr.17: 70384883 - 70398089 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033698.1 NP_001028870.1 3 - 4 658 72
GenBank mRNA BC099837.1 - 3 - 4 658 72

View Full Product Details