The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207590.1 | NP_997473.1 | 5 - 6 | 479 | 74 |
XM_008771854.1 | XP_008770076.1 | 5 - 6 | 498 | 74 | |
XM_008771855.1 | XP_008770077.1 | 3 - 4 | 501 | 74 | |
XM_008771856.1 | XP_008770078.1 | 4 - 5 | 447 | 74 | |
GenBank mRNA | AJ629217.1 | - | 5 - 6 | 479 | 74 |