Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fgl1
Entrez Gene ID
246186
Gene Name
fibrinogen-like 1
Gene Aliases
Frep1, Lfire1
Chromosome Location
Chr.16: 54153086 - 54188120 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC088107.1 - 7 - 8 838 69

View Full Product Details