Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klkb1
Entrez Gene ID
25048
Gene Name
kallikrein B, plasma 1
Gene Aliases
KALP15, Kalp1, Klk3
Chromosome Location
Chr.16: 50151128 - 50175407 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012725.2 NP_036857.2 9 - 10 1089 69
XM_006253121.2 XP_006253183.1 9 - 10 1984 69
XM_008771252.1 XP_008769474.1 7 - 8 844 69
XM_008771253.1 XP_008769475.1 7 - 8 938 69
GenBank mRNA BC089815.1 - 9 - 10 1089 69
M30282.1 - 9 - 10 1052 69
M58590.1 - 9 - 10 1052 69

View Full Product Details