Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spata4
Entrez Gene ID
306441
Gene Name
spermatogenesis associated 4
Gene Aliases
Srg2, Tsarg2
Chromosome Location
Chr.16: 39961160 - 39970529 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002852.1 NP_001002852.1 2 - 3 376 101
XM_006253095.2 XP_006253157.1 2 - 3 381 101
GenBank mRNA AY653229.1 - 2 - 3 376 101

View Full Product Details