The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002852.1 | NP_001002852.1 | 5 - 6 | 842 | 132 |
XM_006253095.2 | XP_006253157.1 | 5 - 6 | 826 | 132 | |
GenBank mRNA | AY653229.1 | - | 5 - 6 | 842 | 132 |