The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014000.1 | NP_001014022.1 | 2 - 3 | 722 | 110 |
GenBank mRNA | BC085832.1 | - | 2 - 3 | 722 | 110 |