Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdh24
Entrez Gene ID
498515
Gene Name
cadherin 24, type 2
Gene Aliases
RGD1560161
Chromosome Location
Chr.15: 33277845 - 33287584 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770708.1 XP_008768930.1 2 - 3 484 55

View Full Product Details