Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pabpn1
Entrez Gene ID
116697
Gene Name
poly(A) binding protein, nuclear 1
Gene Aliases
-
Chromosome Location
Chr.15: 33551157 - 33560251 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135008.1 NP_001128480.1 3 - 4 537 68
XM_008770659.1 XP_008768881.1 4 - 5 2192 68

View Full Product Details