The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135008.1 | NP_001128480.1 | 4 - 5 | 640 | 86 |
XM_008770659.1 | XP_008768881.1 | 5 - 6 | 2295 | 86 |