Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ska3
Entrez Gene ID
361047
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
RGD1307201
Chromosome Location
Chr.15: 38077903 - 38097100 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108379.1 NP_001101849.1 7 - 8 1423 80
XM_006252074.1 XP_006252136.1 7 - 8 1229 80
XM_006252075.1 XP_006252137.1 7 - 8 1229 80
XM_008770758.1 XP_008768980.1 7 - 8 1226 80
GenBank mRNA BC166461.1 - 7 - 8 1200 80

View Full Product Details