Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psme2
Entrez Gene ID
29614
Gene Name
proteasome (prosome, macropain) activator subunit 2
Gene Aliases
-
Chromosome Location
Chr.15: 34265552 - 34269851 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017257.2 NP_058953.1 1 - 2 56 68
GenBank mRNA BC058486.1 - 1 - 2 94 68
BC097331.1 - 1 - 2 56 68
D45250.1 - 1 - 2 106 68
FQ226805.1 - 1 - 2 87 68
FQ227049.1 - 1 - 2 100 68
FQ233763.1 - 1 - 2 82 68

View Full Product Details