The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013995.1 | NP_001014017.1 | 6 - 7 | 730 | 73 |
XM_006251955.1 | XP_006252017.1 | 6 - 7 | 702 | 73 | |
XM_006251956.2 | XP_006252018.1 | 6 - 7 | 881 | 73 | |
GenBank mRNA | BC079109.1 | - | 6 - 7 | 730 | 73 |