Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cideb
Entrez Gene ID
364388
Gene Name
cell death-inducing DFFA-like effector b
Gene Aliases
-
Chromosome Location
Chr.15: 34439844 - 34444244 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108869.1 NP_001102339.1 2 - 3 359 144

View Full Product Details