Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbln3
Entrez Gene ID
501998
Gene Name
cerebellin 3 precursor
Gene Aliases
RGD1563731
Chromosome Location
Chr.15: 34548113 - 34550940 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109330.1 NP_001102800.1 3 - 4 956 82

View Full Product Details