The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012041.1 | NP_001012041.1 | 7 - 8 | 993 | 66 |
XM_006251974.2 | XP_006252036.1 | 7 - 8 | 1121 | 66 | |
XM_006251975.1 | XP_006252037.1 | 7 - 8 | 1065 | 66 | |
XM_008770679.1 | XP_008768901.1 | 7 - 8 | 1106 | 66 | |
GenBank mRNA | BC079454.1 | - | 7 - 8 | 993 | 66 |