The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003711.1 | NP_001003711.1 | 4 - 5 | 1809 | 93 |
XM_008770690.1 | XP_008768912.1 | 5 - 6 | 2607 | 93 | |
GenBank mRNA | AY341260.1 | - | 4 - 5 | 1809 | 93 |