Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serinc5
Entrez Gene ID
170907
Gene Name
serine incorporator 5
Gene Aliases
Tpo1
Chromosome Location
Chr.2: 22195507 - 22310286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ103710.1 - 1 - 2 64 108
L20319.1 - 1 - 2 185 108

View Full Product Details