Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mier3
Entrez Gene ID
310086
Gene Name
mesoderm induction early response 1, family member 3
Gene Aliases
RGD1311241
Chromosome Location
Chr.2: 43266316 - 43295175 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168000.1 NP_001161472.1 2 - 3 276 140
XM_006231924.2 XP_006231986.1 4 - 5 371 140
XM_006231925.2 XP_006231987.2 4 - 5 778 140
GenBank mRNA JQ013732.1 - 2 - 3 1222 140

View Full Product Details