Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dimt1
Entrez Gene ID
294718
Gene Name
DIM1 dimethyladenosine transferase 1 homolog (S. cerevisiae)
Gene Aliases
Dimt1l, RGD1311752
Chromosome Location
Chr.2: 38120558 - 38145747 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106408.2 NP_001099878.2 10 - 11 990 100
XM_006231902.2 XP_006231964.1 10 - 11 1018 100
XM_006231903.2 XP_006231965.1 11 - 12 1201 100
XM_006231905.2 XP_006231967.1 9 - 10 904 100
GenBank mRNA BC158729.1 - 10 - 11 990 100

View Full Product Details