Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd32
Entrez Gene ID
294601
Gene Name
ankyrin repeat domain 32
Gene Aliases
-
Chromosome Location
Chr.2: 4095126 - 4195819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106400.1 NP_001099870.1 17 - 18 2398 114
XM_008760599.1 XP_008758821.1 15 - 16 2040 114

View Full Product Details