Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Papd4
Entrez Gene ID
361878
Gene Name
PAP associated domain containing 4
Gene Aliases
-
Chromosome Location
Chr.2: 22746290 - 22798252 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008372.1 NP_001008373.1 6 - 7 1009 145
XM_006231776.2 XP_006231838.1 6 - 7 999 145
XM_008760639.1 XP_008758861.1 7 - 8 1112 145
GenBank mRNA BC085771.1 - 6 - 7 1009 145

View Full Product Details