Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnpo1
Entrez Gene ID
309126
Gene Name
transportin 1
Gene Aliases
-
Chromosome Location
Chr.2: 29056219 - 29126884 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100692.1 NP_001094162.1 9 - 10 969 79
XM_006231839.2 XP_006231901.1 10 - 11 1560 79
XM_006231840.2 XP_006231902.1 10 - 11 1284 79
XM_008760668.1 XP_008758890.1 10 - 11 1572 79
GenBank mRNA BC090323.1 - 7 - 8 649 79

View Full Product Details