Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lysmd3
Entrez Gene ID
315923
Gene Name
LysM, putative peptidoglycan-binding, domain containing 3
Gene Aliases
RGD1308805
Chromosome Location
Chr.2: 9526213 - 9532370 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009698.1 NP_001009698.1 1 - 2 167 88
GenBank mRNA BC088262.1 - 1 - 2 167 88

View Full Product Details