Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim23
Entrez Gene ID
81002
Gene Name
tripartite motif-containing 23
Gene Aliases
Ard1, Arfd1
Chromosome Location
Chr.2: 34255305 - 34288140 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100637.1 NP_001094107.1 4 - 5 731 68
XM_008760683.1 XP_008758905.1 5 - 6 812 68
GenBank mRNA L04760.1 - 4 - 5 582 68

View Full Product Details