Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cacybp
Entrez Gene ID
289144
Gene Name
calcyclin binding protein
Gene Aliases
-
Chromosome Location
Chr.13: 77948774 - 77959089 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004208.1 NP_001004208.1 4 - 5 592 104
GenBank mRNA BC079007.1 - 4 - 5 592 104

View Full Product Details