Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddx59
Entrez Gene ID
289402
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 59
Gene Aliases
-
Chromosome Location
Chr.13: 53299872 - 53324824 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005535.2 NP_001005535.1 2 - 3 943 77
NM_001190820.1 NP_001177749.1 2 - 3 943 77
GenBank mRNA BC081871.1 - 2 - 3 897 77
CO398251.1 - 1 - 2 291 77

View Full Product Details