The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053560.1 | NP_446012.1 | 9 - 10 | 1067 | 116 |
XM_006249892.2 | XP_006249954.2 | 9 - 10 | 1121 | 116 | |
XM_008769574.1 | XP_008767796.1 | 9 - 10 | 1088 | 116 | |
GenBank mRNA | AF062038.1 | - | 7 - 8 | 948 | 116 |
BC091365.1 | - | 9 - 10 | 1067 | 116 | |
EF467168.1 | - | 7 - 8 | 948 | 116 |