The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107174.1 | NP_001100644.1 | 12 - 13 | 1761 | 63 |
XM_006249775.2 | XP_006249837.1 | 12 - 13 | 1549 | 63 |