Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl11b
Entrez Gene ID
289167
Gene Name
methyltransferase like 11B
Gene Aliases
NTM1B, RGD1564106
Chromosome Location
Chr.13: 81987877 - 82006084 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_222824.7 XP_222824.5 1 - 2 500 79

View Full Product Details