Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl8a
Entrez Gene ID
498234
Gene Name
ADP-ribosylation factor-like 8A
Gene Aliases
RGD1565940
Chromosome Location
Chr.13: 51972974 - 51981976 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109071.1 NP_001102541.1 1 - 2 280 102

View Full Product Details