Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppp1r15b
Entrez Gene ID
304799
Gene Name
protein phosphatase 1, regulatory subunit 15B
Gene Aliases
-
Chromosome Location
Chr.13: 49932405 - 49940961 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107175.1 NP_001100645.1 1 - 2 2282 110

View Full Product Details