Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Suco
Entrez Gene ID
360863
Gene Name
SUN domain containing ossification factor
Gene Aliases
Dd25
Chromosome Location
Chr.13: 79736485 - 79801574 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199403.1 NP_955435.1 10 - 11 1163 133
XM_006250164.2 XP_006250226.1 10 - 11 1618 133
XM_006250166.2 XP_006250228.1 10 - 11 1618 133
XM_008769657.1 XP_008767879.1 10 - 11 1630 133
XM_008769658.1 XP_008767880.1 10 - 11 1630 133
XM_008769659.1 XP_008767881.1 10 - 11 1630 133
GenBank mRNA AJ421447.1 - 10 - 11 1163 133

View Full Product Details