Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March3
Entrez Gene ID
364878
Gene Name
membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.18: 51844980 - 51996984 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007759.1 NP_001007760.1 2 - 3 432 102
XM_006254749.2 XP_006254811.1 2 - 3 396 102
XM_008772157.1 XP_008770379.1 4 - 5 5976 102
XM_008772158.1 XP_008770380.1 3 - 4 3780 102
XM_008772159.1 XP_008770381.1 3 - 4 2592 102
XM_008772160.1 XP_008770382.1 4 - 5 2728 102
GenBank mRNA AB048840.1 - 1 - 2 215 102
BC083738.1 - 2 - 3 432 102

View Full Product Details