Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mex3c
Entrez Gene ID
307271
Gene Name
mex-3 RNA binding family member C
Gene Aliases
Rkhd2
Chromosome Location
Chr.18: 69549937 - 69571736 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107377.1 NP_001100847.1 1 - 2 222 67

View Full Product Details