Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phax
Entrez Gene ID
286917
Gene Name
phosphorylated adaptor for RNA export
Gene Aliases
Rnuxa
Chromosome Location
Chr.18: 51662294 - 51678981 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173133.2 NP_775156.2 4 - 5 907 117
GenBank mRNA BC086410.1 - 4 - 5 877 117
BC097932.1 - 4 - 5 883 117
FQ226770.1 - 3 - 4 455 117
X67877.1 - 4 - 5 906 117

View Full Product Details