The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109539.1 | NP_001103009.1 | 4 - 5 | 420 | 82 |
XM_008772175.1 | XP_008770397.1 | 4 - 5 | 342 | 82 |