Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdo1
Entrez Gene ID
81718
Gene Name
cysteine dioxygenase type 1
Gene Aliases
-
Chromosome Location
Chr.18: 40702027 - 40716902 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052809.1 NP_434696.1 4 - 5 760 77
GenBank mRNA BC070509.1 - 4 - 5 751 77
FQ219282.1 - 3 - 4 397 77
M35266.1 - 4 - 5 760 77

View Full Product Details