The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038495.1 | NP_001033584.1 | 3 - 4 | 384 | 128 |
GenBank mRNA | BC087139.1 | - | 3 - 4 | 367 | 128 |
BC110056.1 | - | 3 - 4 | 384 | 128 |