Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grpel2
Entrez Gene ID
688777
Gene Name
GrpE-like 2, mitochondrial
Gene Aliases
Afap1l1
Chromosome Location
Chr.18: 57037995 - 57050234 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109513.1 NP_001102983.1 2 - 3 254 143

View Full Product Details