The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107884.1 | NP_001101354.1 | 7 - 8 | 946 | 91 |
XM_006237246.2 | XP_006237308.1 | 7 - 8 | 1421 | 91 |