Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcp1b
Entrez Gene ID
500305
Gene Name
decapping mRNA 1B
Gene Aliases
RGD1562214
Chromosome Location
Chr.4: 151249655 - 151287018 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_575654.6 XP_575654.3 7 - 8 1928 80

View Full Product Details