Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyrm5
Entrez Gene ID
100361188
Gene Name
LYR motif containing 5
Gene Aliases
-
Chromosome Location
Chr.4: 179477081 - 179483434 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729439.4 XP_002729485.1 2 - 3 205 139
XM_006237672.2 XP_006237734.1 3 - 4 323 139
XM_006237673.2 XP_006237735.1 1 - 2 80 139

View Full Product Details