The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212513.2 | NP_997678.2 | 5 - 6 | 1162 | 91 |
XM_006237348.2 | XP_006237410.1 | 5 - 6 | 1387 | 91 | |
XM_008763282.1 | XP_008761504.1 | 6 - 7 | 1268 | 91 | |
XM_008763283.1 | XP_008761505.1 | 4 - 5 | 912 | 91 | |
GenBank mRNA | BC091201.1 | - | 5 - 6 | 1162 | 91 |
DQ438937.1 | - | 5 - 6 | 1040 | 91 |