Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan9
Entrez Gene ID
312728
Gene Name
tetraspanin 9
Gene Aliases
RGD1304740
Chromosome Location
Chr.4: 160530726 - 160725048 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107890.1 NP_001101360.1 1 - 2 434 137

View Full Product Details