Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Styk1
Entrez Gene ID
500340
Gene Name
serine/threonine/tyrosine kinase 1
Gene Aliases
RGD1564211
Chromosome Location
Chr.4: 165528027 - 165544659 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763441.1 XP_008761663.1 2 - 3 774 117
XM_008763442.1 XP_008761664.1 4 - 5 762 117
XM_008763443.1 XP_008761665.1 5 - 6 1517 117

View Full Product Details