Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcat1
Entrez Gene ID
29592
Gene Name
branched chain amino acid transaminase 1, cytosolic
Gene Aliases
Bcatc
Chromosome Location
Chr.4: 179259305 - 179340021 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017253.2 NP_058949.1 3 - 4 445 63
XM_008763370.1 XP_008761592.1 3 - 4 458 63
XM_008763371.1 XP_008761593.1 3 - 4 708 63
XM_008763372.1 XP_008761594.1 4 - 5 534 63
XM_008763373.1 XP_008761595.1 3 - 4 637 63
GenBank mRNA AJ278701.1 - 3 - 4 439 63
BC087710.1 - 3 - 4 445 63
U35774.1 - 3 - 4 411 63

View Full Product Details